3'-Deoxyadenosine (cordycepine) 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3'-Deoxyadenosine, (2R,3R,5S)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3-ol, Cordycepin
J65366-MB
J65366-MC
10814-388EA
177
USD
10814-388
10814-390
3'-Deoxyadenosine (cordycepine) 98%
3'-Deoxyadenosine (cordycepine)
Purity: 98%
CAS No.: 73-03-0
Molecular Formula: C10H13N5O3
Formula Weight: 251.24
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Beilstein Reference: 35194
Merck Reference: 14,2529
UN No.: 2811
MDL No.: MFCD00037998
Formula:
C₁₀H₁₃N₅O₃ MW: 251.24 Storage Temperature: Freezer |
MDL Number:
MFCD00037998 CAS Number: 73-03-0 EINECS: 200-791-4 UN: 2811 ADR: "6.1",III Merck Index: 14,2529 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...