Atropine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(+/-)-Hyoscyamine, Tropine DL-tropate
J64677-14
J64677-03
10814-336EA
416
USD
10814-336
10815-312
Atropine 99%
Atropine
Reduces brain levels of acetylcholine
Purity: 99%
CAS No.: 51-55-8
Molecular Formula: C17H23NO3
Formula Weight: 289.37
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: White
Beilstein Reference: 91260
Merck Reference: 14,875
UN No.: 1544
MDL No.: MFCD00022622
Reduces brain levels of acetylcholine
Formula:
C₁₇H₂₃NO₃ MW: 289.38 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00022622 CAS Number: 51-55-8 EINECS: 200-104-8 UN: 1544 ADR: "6.1",III Merck Index: 14,875 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...