Bleomycin sulfate (mixture)
Supplier: Thermo Scientific Chemicals
|
Danger
|
J60727-MA
J60727-MCR
10814-234EA
431.03
USD
10814-234
10814-236
Bleomycin sulfate (mixture)
Bleomycin sulfate (mixture)
A glycopeptide antibiotic. Inhibits DNA synthesis and causes breaks in DNA strands
White to off-white powder
It is capable of inducing oxidative cleavage of the DNA backbone, leading to apoptosis and also degrades RNA, and has been reported to display selectivity for RNA over DNA. Bleomycin acts as a sclerosing agent.
Soluble in water or DMSO
Precaución: Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Formula:
C₅₅H₈₄N₁₇O₂₁S₃·xH₂SO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00070310 CAS Number: 9041-93-4 EINECS: 232-925-2 Merck Index: 14,1318 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...