4-Aminoquinazoline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Quinazolin-4-amine, Quinazolin-4-ylamine
H66948-03
H66948-06
AAH66948-03EA
122
USD
AAH66948-03
AAH66948-06
4-Aminoquinazoline 97%
4-Aminoquinazoline
Purity: 97%
CAS No.: 15018-66-3
Molecular Formula: C8H7N3
Formula Weight: 145.06
Physical Form: Solid
Formula:
C₈H₇N₃ MW: 145.06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00092016 CAS Number: 15018-66-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...