5-Cyano-2-methoxybenzaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Formyl-4-methoxybenzonitrile
H66690-03
H66690-MD
AAH66690-03EA
163
USD
AAH66690-03
AAH66690-MD
5-Cyano-2-methoxybenzaldehyde 97%
5-Cyano-2-methoxybenzaldehyde
Purity: 97%
CAS No.: 21962-53-8
Molecular Formula: C9H7NO2
Formula Weight: 161.16
Physical Form: Solid
Sensitivity: Air Sensitive
Appearance: Pale yellow to dark yello
Melting Point: 115-120°
UN No.: 3439
MDL No.: MFCD13195275
Formula:
C₉H₇NO₂ MW: 161.16 g/mol Melting Pt: 115…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD13195275 CAS Number: 21962-53-8 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...