Methyl oxindole-7-carboxylate 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 2-oxoindoline-7-carboxylate

H66091-03 H66091-MD
AAH66091-03EA 391 USD
AAH66091-03 AAH66091-MD
Methyl oxindole-7-carboxylate 96%
Methyl oxindole-7-carboxylate

Purity: 96% CAS No.: 380427-39-4 Molecular Formula: C10H9NO3 Formula Weight: 191.19 Physical Form: Solid

Formula: C₁₀H₉NO₃
MW: 191.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179610
CAS Number: 380427-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR