3-Bromo-4-nitroindole 97%

Supplier: Thermo Scientific Chemicals
Warning

H63478-03 H63478-06 H63478-MD
AAH63478-03EA 129 USD
AAH63478-03 AAH63478-06 AAH63478-MD
3-Bromo-4-nitroindole 97%
3-Bromo-4-nitroindole

Purity: 97% CAS No.: 126807-08-7 Molecular Formula: C8H5BrN2O2 Formula Weight: 241.04 Storage Temperature: 2°C to 8°C Physical Form: Solid

Formula: C₈H₅BrN₂O₂
Storage Temperature: Ambient
CAS Number: 126807-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR