3-Cyano-4-methylaniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Amino-2-methylbenzonitrile
H61422-06
H61422-14
AAH61422-06EA
71.9
USD
AAH61422-06
AAH61422-14
3-Cyano-4-methylaniline 97%
3-Cyano-4-methylaniline
Purity: 97%
CAS No.: 50670-64-9
Molecular Formula: C8H8N2
Formula Weight: 132.16
Physical Form: Solid
Melting Point: 88-91°
UN No.: 3439
MDL No.: MFCD00017608
Formula:
C₈H₈N₂ MW: 132.16 g/mol Melting Pt: 88…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017608 CAS Number: 50670-64-9 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...