5-Bromo-2-(4-morpholinyl)benzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

H61376-03 H61376-MD
AAH61376-03EA 278 USD
AAH61376-03 AAH61376-MD
5-Bromo-2-(4-morpholinyl)benzonitrile 98%
5-Bromo-2-(4-morpholinyl)benzonitrile

Purity: 98% CAS No.: 1105665-08-4 Molecular Formula: C11H11BrN2O Formula Weight: 267.13 Physical Form: Solid UN No.: 3439

Formula: C₁₁H₁₁BrN₂O
MW: 267.126 g/mol
Storage Temperature: Ambient
CAS Number: 1105665-08-4
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR