(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-(1-Ethylpyrrolidin-2-yl)methanamine, (S)-2-Aminomethyl-1-ethylpyrrolidine, (S)-1-ethyl-2-aminomethylpyrrolidine

H57137-03 H57137-06
AAH57137-03EA 53.7 USD
AAH57137-03 AAH57137-06
(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine 95%
(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine

Purity: 95% CAS No.: 22795-99-9 Molecular Formula: C7H16N2 Formula Weight: 128.22 Flash Point: 57°(135°F) Refractive Index: 1.4660 Boiling Point: 50-54° Optical Rotation: -95° (c=1 in methanol) UN No.: 2733 MDL No.: MFCD00191371

Formula: C₇H₁₆N₂
MW: 128.22 g/mol
Boiling Pt: 50…54 °C
Density: 0.919
Flash Pt: 57 °C (135 °F)
Storage Temperature: Ambient
MDL Number: MFCD00191371
CAS Number: 22795-99-9
UN: 2734
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR