Phenyl methanesulfonate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methanesulphonic acid Phenyl Ester
H55995-03
H55995-06
AAH55995-03EA
20.8
USD
AAH55995-03
AAH55995-06
AAH55995-14
Phenyl methanesulfonate 98%
Phenyl methanesulfonate
Purity: 98%
CAS No.: 16156-59-5
Molecular Formula: C7H8O3S
Formula Weight: 172.20
Physical Form: Solid
Appearance: Pale brown
Melting Point: 58-61°
Boiling Point: 279°
MDL No.: MFCD00095143
Formula:
C₇H₈O₃S MW: 172.20 g/mol Boiling Pt: 279 °C Melting Pt: 58…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00095143 CAS Number: 16156-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...