4-Chlorophenylboronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(4-Chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Chlorophenylboronic acid Pinacol ester, 4-Chlorobenzeneboronic acid pinacol ester, 1-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

H55984-03 H55984-06 H55984-14
AAH55984-03EA 30.6 USD
AAH55984-03 AAH55984-06 AAH55984-14
4-Chlorophenylboronic acid pinacol ester 97%
4-Chlorophenylboronic acid pinacol ester

Purity: 97% CAS No.: 195062-61-4 Molecular Formula: C12H16BClO2 Formula Weight: 238.52 Flash Point: 110°(230°F) Physical Form: Solid Melting Point: 50-55° MDL No.: MFCD05663875

Formula: C₁₂H₁₆BClO₂
MW: 238.52 g/mol
Melting Pt: 50…55 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD05663875
CAS Number: 195062-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR