1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DiI, DiI perchlorate

H55957-MC H55957-ME
AAH55957-MCEA 206 USD
AAH55957-MC AAH55957-ME
1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate 97%
1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate

Purity: 97% CAS No.: 41085-99-8 Molecular Formula: C59H97ClN2O4 Formula Weight: 933.88 Melting Point: 68° dec. Beilstein Reference: 8251870

Formula: C₅₉H₉₇ClN₂O₄
MW: 933.885 g/mol
Melting Pt: 68 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00142354
CAS Number: 41085-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR