1-(1-Naphthyl)-2-(trimethylsilyl)acetylene 97%

Supplier: Thermo Scientific Chemicals

H55953-03 H55953-06 H55953-14
AAH55953-03EA 37.5 USD
AAH55953-03 AAH55953-06 AAH55953-14
1-(1-Naphthyl)-2-(trimethylsilyl)acetylene 97%
1-(1-Naphthyl)-2-(trimethylsilyl)acetylene

Purity: 97% CAS No.: 104784-51-2 Molecular Formula: C15H16Si Formula Weight: 224.38 Flash Point: 110°(230°F) Physical Form: Solid Density: 0.968 Refractive Index: 1.5950 Melting Point: 41-43° Boiling Point: 292-293° MDL No.: MFCD04039886

Formula: C₁₅H₁₆Si
Boiling Pt: 292…293 °C
Melting Pt: 41…43 °C
Density: 0.968
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD04039886
CAS Number: 104784-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR