4-tert-Butylphenyl glycidyl ether 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55945-06
H55945-14
AAH55945-06EA
44.3
USD
AAH55945-06
AAH55945-14
4-tert-Butylphenyl glycidyl ether 95%
4-tert-Butylphenyl glycidyl ether
Used in paint industry and serves as organic coating in paint, boat paint, building coating material and also as an epoxy reactive diluent.
Purity: 95%
CAS No.: 3101-60-8
Molecular Formula: C13H18O2
Formula Weight: 206.28
Flash Point: 102°(216°F)
Physical Form: Liquid
Density: 1.038
Refractive Index: 1.5150
Appearance: Colorless
Boiling Point: 165-170°/14mm
MDL No.: MFCD00005136
Formula:
C₁₃H₁₈O₂ Boiling Pt: 165…170 °C/14 mm Density: 1.038 Flash Pt: 102 °C(216 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005136 CAS Number: 3101-60-8 EINECS: 221-453-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...