2-[(Trimethylsilyl)ethynyl]aniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55930-03
H55930-06
H55930-14
AAH55930-03EA
57.8
USD
AAH55930-03
AAH55930-06
AAH55930-14
2-[(Trimethylsilyl)ethynyl]aniline 97%
2-[(Trimethylsilyl)ethynyl]aniline
Purity: 97%
CAS No.: 103529-16-4
Molecular Formula: C11H15NSi
Formula Weight: 189.33
Flash Point: 110°(230°F)
Physical Form: Liquid
Density: 0.94
Refractive Index: 1.5640
Appearance: Colorless to pale yellow
Boiling Point: 120°/0.1mm
MDL No.: MFCD02093646
Formula:
C₁₁H₁₅NSi MW: 189.333 g/mol Boiling Pt: 120 °C/0.1 mm Density: 0.94 Flash Pt: 110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD02093646 CAS Number: 103529-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...