(3-Benzyloxypropyl)triphenylphosphonium bromide 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
[3-(Benzyloxy)propyl]triphenylphosphonium Bromide
H55754-06
H55754-14
AAH55754-03EA
43.41
USD
AAH55754-03
AAH55754-06
AAH55754-14
(3-Benzyloxypropyl)triphenylphosphonium bromide 97+%
(3-Benzyloxypropyl)triphenylphosphonium bromide
Purity: 98%
CAS No.: 54314-85-1
Molecular Formula: C28H28BrOP
Formula Weight: 491.41
Physical Form: Powder
Appearance: White to off-white
Melting Point: 153-154°
MDL No.: MFCD00191781
Formula:
C₂₈H₂₈BrOP MW: 491.41 g/mol Melting Pt: 153…154 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191781 CAS Number: 54314-85-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...