4-N-Boc-aminophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: tert-Butyl (4-hydroxyphenyl)carbamate, 4-(Boc-amino)phenol

H55645-06 H55645-14
AAH55645-06EA 62 USD
AAH55645-06 AAH55645-14
4-N-Boc-aminophenol 97%
4-N-Boc-aminophenol

Purity: 97% CAS No.: 54840-15-2 Molecular Formula: C11H15NO3 Formula Weight: 209.25 Melting Point: 143-147° MDL No.: MFCD00226573

Formula: C₁₁H₁₅NO₃
MW: 209.245 g/mol
Melting Pt: 143…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00226573
CAS Number: 54840-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR