(6-Bromohexyloxy)tert-butyldimethylsilane 99%

Supplier: Thermo Scientific Chemicals
Warning

H55581-AA H55581-AC
AAH55581-AAEA 101 USD
AAH55581-AA AAH55581-AC
(6-Bromohexyloxy)tert-butyldimethylsilane 99%
(6-Bromohexyloxy)tert-butyldimethylsilane

Purity: 99% CAS No.: 129368-70-3 Molecular Formula: C12H27BrOSi Formula Weight: 295.34 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.053 Refractive Index: 1.4570 Appearance: Colorless Boiling Point: 276° MDL No.: MFCD01863708

Formula: C₁₂H₂₇BrOSi
Boiling Pt: 276 °C
Density: 1.053
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01863708
CAS Number: 129368-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR