Bis(2,2,2-trifluoroethyl)phosphite 90%, Technical Grade
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Phosphorous Acid Bis(2,2,2-trifluoroethyl) Ester, Bis(2,2,2-trifluoroethyl) Phosphonate, Phosphonic Acid Bis(2,2,2-trifluoroethyl) Ester
H55562-03
H55562-06
AAH55562-03EA
24.5
USD
AAH55562-03
AAH55562-06
Bis(2,2,2-trifluoroethyl)phosphite 90%, Technical Grade
Bis(2,2,2-trifluoroethyl)phosphite
Purity: 90%
CAS No.: 92466-70-1
Molecular Formula: C4F6H5O3P
Formula Weight: 246.05
Flash Point: 76°(169°F)
Physical Form: Liquid
Density: 1.545
Refractive Index: 1.3320
Boiling Point: 43-44°/2mm
MDL No.: MFCD00063314
Formula:
C₄H₅F₆O₃P MW: 246.05 g/mol Boiling Pt: 43…44 °C/2 mm Density: 1.545 Flash Pt: 76 °C(169 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00063314 CAS Number: 92466-70-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...