1-Boc-4-(3-hydroxypropyl)piperazine 97%

Supplier: Thermo Scientific Chemicals
Danger

H55547-06 H55547-14
AAH55547-03EA 25.54 USD
AAH55547-03 AAH55547-06 AAH55547-14
1-Boc-4-(3-hydroxypropyl)piperazine 97%
1-Boc-4-(3-hydroxypropyl)piperazine

Purity: 97% CAS No.: 132710-90-8 Molecular Formula: C12H24N2O3 Formula Weight: 244.33 Physical Form: Solid Melting Point: 73-77° MDL No.: MFCD06798090

Formula: C₁₂H₂₄N₂O₃
MW: 244.335 g/mol
Melting Pt: 73…77 °C
Storage Temperature: Ambient
MDL Number: MFCD06798090
CAS Number: 132710-90-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR