Diphenylborinic anhydride 95%
Supplier: Thermo Scientific Chemicals
H55509-03
H55509-06
H55509-MD
AAH55509-03EA
235
USD
AAH55509-03
AAH55509-06
AAH55509-MD
Diphenylborinic anhydride 95%
Diphenylborinic anhydride
Purity: 95%
CAS No.: 4426-21-5
Molecular Formula: C24H20B2O
Formula Weight: 346.04
Physical Form: Powder
Sensitivity: Moisture Sensitive
Appearance: White to yellow
Melting Point: 135-140°
Beilstein Reference: 2950331
MDL No.: MFCD00042663
Formula:
C₂₄H₂₀B₂O Melting Pt: 135…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042663 CAS Number: 4426-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...