4-Hydroxy-4-phenylpiperidine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Phenyl-4-piperidinol

H55418-03 H55418-06
AAH55418-03EA 64.3 USD
AAH55418-03 AAH55418-06 AAH55418-14
4-Hydroxy-4-phenylpiperidine 99%
4-Hydroxy-4-phenylpiperidine

Purity: 99% CAS No.: 40807-61-2 Molecular Formula: C11H15NO Formula Weight: 177.25 Physical Form: Solid Appearance: Pale brown Melting Point: 157-161° Beilstein Reference: 141577 MDL No.: MFCD00006000

Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
Melting Pt: 157…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00006000
CAS Number: 40807-61-2
EINECS: 255-089-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR