Bis(2-aminophenyl) ether 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2'-Oxydianiline

H55396-03 H55396-06 H55396-MD
AAH55396-03EA 83.3 USD
AAH55396-03 AAH55396-06 AAH55396-MD
Bis(2-aminophenyl) ether 98%
Bis(2-aminophenyl)ether

Purity: 98% CAS No.: 24878-25-9 Molecular Formula: C12H12N2O Formula Weight: 200.24 Physical Form: Powder Appearance: White to pale brown Melting Point: 61-64° MDL No.: MFCD00792533

Formula: C₁₂H₁₂N₂O
MW: 200.241 g/mol
Melting Pt: 61…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00792533
CAS Number: 24878-25-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR