9,9-Di-n-octylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
2,2'-(9,9-Dioctyl-9H-fluorene-2,7-diyl)bis(1,3,2-dioxaborinane)
H55392-03
H55392-MD
AAH55392-03EA
98
USD
AAH55392-03
AAH55392-MD
9,9-Di-n-octylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester 97%
9,9-Di-n-octylfluorene-2,7-diboronic acid bis(1,3-propanediol) ester
Purity: 97%
CAS No.: 317802-08-7
Molecular Formula: C35H52B2O4
Formula Weight: 558.42
Physical Form: Solid
Formula:
C₃₅H₅₂B₂O₄ MW: 558.417 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03701607 CAS Number: 317802-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...