2-Amino-7-bromofluorene 90%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

H55370-03 H55370-06
AAH55370-03EA 72.2 USD
AAH55370-03 AAH55370-06 AAH55370-MD
2-Amino-7-bromofluorene 90%, Technical Grade
2-Amino-7-bromofluorene

Purity: 90% CAS No.: 6638-60-4 Molecular Formula: C13H10BrN Formula Weight: 260.13 Melting Point: 148-149° MDL No.: MFCD00001127

Formula: C₁₃H₁₀BrN
Melting Pt: 148…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00001127
CAS Number: 6638-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR