4-Acetyl-N-methylaniline 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-(4-(Methylamino)phenyl)ethanone, 4'-(Methylamino)acetophenone
H55301-03
H55301-MD
AAH55301-03EA
104
USD
AAH55301-03
AAH55301-06
AAH55301-MD
4-Acetyl-N-methylaniline 97%
4-Acetyl-N-methylaniline
Purity: 97%
CAS No.: 17687-47-7
Molecular Formula: C9H11NO
Formula Weight: 149.19
Physical Form: Solid
Appearance: White to pale brown
Melting Point: 103-107°
MDL No.: MFCD09261160
Formula:
C₉H₁₁NO MW: 149.193 g/mol Melting Pt: 103…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD09261160 CAS Number: 17687-47-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...