1-(3-Fluorobenzyl)piperazin 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55295-03
H55295-06
AAH55295-03EA
50.5
USD
AAH55295-03
AAH55295-06
AAH55295-14
1-(3-Fluorobenzyl)piperazin 97%
1-(3-Fluorobenzyl)piperazin
Purity: 97%
CAS No.: 55513-19-4
Molecular Formula: C11H15FN2
Formula Weight: 194.25
Flash Point:
Formula:
C₁₁H₁₅FN₂ MW: 194.25 g/mol Boiling Pt: 281 °C Density: 1.106 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD02242870 CAS Number: 55513-19-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...