4-Bromo-N-methylaniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55273-03
H55273-06
AAH55273-03EA
47.1
USD
AAH55273-03
AAH55273-06
AAH55273-14
4-Bromo-N-methylaniline 97%
4-Bromo-N-methylaniline
Purity: 97%
CAS No.: 6911-87-1
Molecular Formula: C7H8BrN
Formula Weight: 186.05
Physical Form: Liquid
Density: 1.482
Refractive Index: 1.6130
Boiling Point: 259-260°
MDL No.: MFCD03030391
Formula:
C₇H₈BrN MW: 186.05 g/mol Boiling Pt: 259…260 °C Density: 1.482 Storage Temperature: Ambient |
MDL Number:
MFCD03030391 CAS Number: 6911-87-1 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...