N-Boc-4-bromoaniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromoaniline, N-BOC protected, tert-Butyl (4-bromophenyl)carbamate
H55226-06
H55226-14
AAH55226-03EA
18.65
USD
AAH55226-03
AAH55226-06
AAH55226-14
N-Boc-4-bromoaniline 97%
N-Boc-4-bromoaniline
Purity: 97%
CAS No.: 131818-17-2
Molecular Formula: C11H14BrNO2
Formula Weight: 272.14
Physical Form: Solid
Appearance: White to pale brown
Melting Point: 103-106°
MDL No.: MFCD01006612
Formula:
C₁₁H₁₄BrNO₂ Melting Pt: 103…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD01006612 CAS Number: 131818-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...