1-(Cyclopropylcarbonyl)piperazine 97%

Supplier: Thermo Scientific Chemicals
Danger

H55219-03 H55219-06 H55219-MD
AAH55219-03EA 89.1 USD
AAH55219-03 AAH55219-06 AAH55219-MD
1-(Cyclopropylcarbonyl)piperazine 97%
1-(Cyclopropylcarbonyl)piperazine

Purity: 97% CAS No.: 59878-57-8 Molecular Formula: C8H14N2O Formula Weight: 154.21 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.087 Refractive Index: 1.5240 Appearance: Colorless to pale brown Boiling Point: 120°/15mm MDL No.: MFCD06369645

Formula: C₈H₁₄N₂O
MW: 154.213 g/mol
Boiling Pt: 120 °C/15 mm
Density: 1.087
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD06369645
CAS Number: 59878-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR