N-Methyl-3-nitroaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

H55196-03 H55196-MD
AAH55196-03EA 199 USD
AAH55196-03 AAH55196-06 AAH55196-MD
N-Methyl-3-nitroaniline 97%
N-Methyl-3-nitroaniline

Purity: 97% CAS No.: 619-26-1 Molecular Formula: C7H8N2O2 Formula Weight: 152.15 Physical Form: Powder Appearance: White to brown Melting Point: 64-68° MDL No.: MFCD00963641

Formula: C₇H₈N₂O₂
MW: 152.153 g/mol
Melting Pt: 64…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00963641
CAS Number: 619-26-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR