2,7-Dinitro-9-fluorenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55176-06
H55176-14
H55176-22
AAH55176-06EA
55.9
USD
AAH55176-06
AAH55176-14
AAH55176-22
2,7-Dinitro-9-fluorenone 97%
2,7-Dinitro-9-fluorenone
Purity: 97%
CAS No.: 31551-45-8
Molecular Formula: C13H6N2O5
Formula Weight: 270.20
Physical Form: Powder
Appearance: Yellow to orange
Melting Point: 292-295°
MDL No.: MFCD00001153
Formula:
C₁₃H₆N₂O₅ MW: 270.2 g/mol Melting Pt: 292…295 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001153 CAS Number: 31551-45-8 EINECS: 250-695-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...