2-Bromo-N-methylaniline 95%

Supplier: Thermo Scientific Chemicals
Warning

H55166-03 H55166-06 H55166-MD
AAH55166-03EA 112 USD
AAH55166-03 AAH55166-06 AAH55166-MD
2-Bromo-N-methylaniline 95%
2-Bromo-N-methylaniline

Purity: 95% CAS No.: 6832-87-7 Molecular Formula: C7H8BrN Formula Weight: 186.05 Flash Point: 103°(217°F) Physical Form: Liquid Density: 1.589 Refractive Index: 1.6070 Appearance: Brown Boiling Point: 107-109°/12mm MDL No.: MFCD06798064

Formula: C₇H₈BrN
MW: 186.05 g/mol
Boiling Pt: 107…109 °C/12 mm
Density: 1.589
Flash Pt: 103 °C(217 °F)
Storage Temperature: Ambient
MDL Number: MFCD06798064
CAS Number: 6832-87-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR