6-Fluoroquinoline 97%

Supplier: Thermo Scientific Chemicals
Warning

H35900-03 H35900-MD
AAH35900-03EA 70.2 USD
AAH35900-03 AAH35900-06 AAH35900-MD
6-Fluoroquinoline 97%
6-Fluoroquinoline

Purity: 97% CAS No.: 396-30-5 Molecular Formula: C9H6FN Formula Weight: 147.15 MDL No.: MFCD01685512

Formula: C₉H₆FN
MW: 147.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01685512
CAS Number: 396-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR