(Iodomethyl)cyclopropane 95%

Supplier: Thermo Scientific Chemicals
Danger

H35309-03 H35309-MD
AAH35309-03EA 186 USD
AAH35309-03 AAH35309-MD
(Iodomethyl)cyclopropane 95%
(Iodomethyl)cyclopropane

Purity: 97% CAS No.: 33574-02-6 Molecular Formula: C4H7I Formula Weight: 182.00 MDL No.: MFCD06410734

Formula: C₄H₇I
MW: 182.00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06410734
CAS Number: 33574-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR