(S)-(+)-1-Bromo-2-methylbutane 97% stabilised

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-(+)-2-Methylbutyl bromide

Stabilization: Stabilised with potassium carbonate

H27078-06 H27078-14
AAH27078-06EA 164.47 USD
AAH27078-06 AAH27078-14
(S)-(+)-1-Bromo-2-methylbutane 97% stabilised
(S)-(+)-1-Bromo-2-methylbutane

Purity: 97% CAS No.: 534-00-9 Molecular Formula: C5H11Br Formula Weight: 151.05 Flash Point: 22°(79°F) Refractive Index: 1.4455 Boiling Point: 121-122° Beilstein Reference: 1718815 Merck Reference: 14,601 Optical Rotation: 4.5° (neat) UN No.: 1993 MDL No.: MFCD00078662

Formula: C₅H₁₁Br
Boiling Pt: 121…122 °C
Density: 1.223
Flash Pt: 22 °C (79 °F)
Storage Temperature: Ambient
MDL Number: MFCD00078662
CAS Number: 534-00-9
EINECS: 208-583-5
UN: 1993
ADR: 3,III
Merck Index: 14,601

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR