Borane-d3 1 M in THF

Supplier: Thermo Scientific Chemicals
Danger

089121-AD 089121-AK
AA89121-ADEA 527 USD
AA89121-AD AA89121-AK
Borane-d3 1 M in THF
Borane-d3

CAS No.: 13763-62-7 Molecular Formula: BD3/(CH2)4O Formula Weight: 16.84 as BD3 Storage Temperature: 2°C to 8°C Flash Point: -21°(-6°F) Physical Form: Liquid Density: 0.876 Sensitivity: Air and Moisture Sensitive Appearance: Colorless Odor: Ether-like UN No.: 3148 MDL No.: MFCD08705283

Formula: BH₃
Density: 0.876
Flash Pt: –21 °C (–6 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD08705283
CAS Number: 13763-62-7
UN: 3148
ADR: 4.3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR