Sklar Black™ Cottle Septal Speculum, OR Grade, Sklar
Supplier: Sklar
41-1198 41-1200
10199-398EA 655.73 USD
10199-398 10199-400
Sklar Black™ Cottle Septal Speculum, OR Grade, Sklar
Specula
Cottle nasal speculum is used in laser procedures of the septum.
  • Stainless steel with reduced reflectivity polymer coating
Sklar Black™ laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff.
All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity and sensitization regulatory.
Certificaciones: Meets ISO 10993 requirements.