Sklar Black™ Bozeman Forceps, OR Grade, Sklar
Supplier: Sklar
90-2340 90-2342
10199-120EA 453.05 USD
10199-120 10199-122
Sklar Black™ Bozeman Forceps, OR Grade, Sklar
Forceps
Sklar Black™ Bozeman Forceps are a multi-functional instrument commonly used in gynecological procedures
They are used for tissue removal in dilation and curettage procedures, intrauterine device retrieval, and packing the uterine cavity. The shanks of this instrument are slightly curved and therefore provide greater visibility when used in the uterine cavity. The jaws are available either curved or straight. Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. All Sklar Black™ coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Black™ laser coatings are guaranteed for one year against manufacturing defects.