Anti-RORC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 600380]

Supplier: R&D Systems
IC6006P-025 IC6006P-100
102884-642EA 302.25 USD
102884-642 102884-644
Anti-RORC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 600380]
Antibodies
Type: Primary
Antigen: RORC
Clonality: Monoclonal
Clone: 600380
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR