Anti-CCR8 Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1055c]

Supplier: R&D Systems
FAB8324P-025 FAB8324P-100
102883-934EA 229.94 USD
102883-934 102883-936
Anti-CCR8 Rabbit Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1055c]
Antibodies
Type: Primary
Antigen: CCR8
Clonality: Monoclonal
Clone: 1055c
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR