Anti-CCR6 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 876515]

Supplier: R&D Systems
FAB8320P-025 FAB8320P-100
102883-926EA 244.4 USD
102883-926 102883-928
Anti-CCR6 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 876515]
Antibodies
Type: Primary
Antigen: CCR6
Clonality: Monoclonal
Clone: 876515
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR