Anti-CCR3 Rat Monoclonal Antibody (Fluorescein) [clone: 83101]

Supplier: R&D Systems
FAB729F-025 FAB729F-100
102883-654EA 201.02 USD
102883-654 102883-656
Anti-CCR3 Rat Monoclonal Antibody (Fluorescein) [clone: 83101]
Antibodies
Type: Primary
Antigen: CCR3
Clonality: Monoclonal
Clone: 83101
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR