Anti-KLRC1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 705829]

Supplier: R&D Systems
FAB6867P-025 FAB6867P-100
102883-434EA 172.09 USD
102883-434 102883-436
Anti-KLRC1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 705829]
Antibodies
Type: Primary
Antigen: KLRC1
Clonality: Monoclonal
Clone: 705829
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR