Anti-CCR1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 643854]

Supplier: R&D Systems
FAB5986P-025 FAB5986P-100
102883-158EA 258.86 USD
102883-158 102883-160
Anti-CCR1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 643854]
Antibodies
Type: Primary
Antigen: CCR1
Clonality: Monoclonal
Clone: 643854
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR