Anti-CX3CR1 Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R&D Systems
FAB5825P-025 FAB5825P-100
102883-106EA 273.32 USD
102883-106 102883-108
Anti-CX3CR1 Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: CX3CR1
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR