Anti-CCR2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 475301]

Supplier: R&D Systems
FAB5538P-025 FAB5538P-100
102883-000EA 258.86 USD
102883-000 102883-002
Anti-CCR2 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 475301]
Antibodies
Type: Primary
Antigen: CCR2
Clonality: Monoclonal
Clone: 475301
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR