Anti-CCR2 Rat Monoclonal Antibody (Fluorescein) [clone: 475301]

Supplier: R&D Systems
FAB5538F-025 FAB5538F-100
102882-992EA 229.94 USD
102882-992 102882-994
Anti-CCR2 Rat Monoclonal Antibody (Fluorescein) [clone: 475301]
Antibodies
Type: Primary
Antigen: CCR2
Clonality: Monoclonal
Clone: 475301
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR