Anti-CXCR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 42705]

Supplier: R&D Systems
FAB330P-100 FAB330P-025
102882-250EA 562.55 USD
102882-250 102882-464
Anti-CXCR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 42705]
Antibodies
Type: Primary
Antigen: CXCR1
Clonality: Monoclonal
Clone: 42705
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR